missing translation for 'onlineSavingsMsg'
Learn More

Erbium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific Chemicals

Chemical Identifiers

CAS 14692-15-0
Molecular Formula Cl3ErH12O18
Molecular Weight (g/mol) 573.687
MDL Number MFCD00150232
InChI Key JZRNHNTYRRYOCR-UHFFFAOYSA-K
Synonym erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
PubChem CID 20038200
IUPAC Name erbium(3+);triperchlorate;hexahydrate
SMILES O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11480301
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040576.NA
∼2 g N/A
N/A
11490301
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040576.NC
10 g N/A
N/A
11400311
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040576.NE
∼50 g N/A
N/A
Description

Description

Erbium(III) perchlorate hexahydrate is used in nuclear technology for neutron-absorbing control rods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

14692-15-0
573.687
JZRNHNTYRRYOCR-UHFFFAOYSA-K
20038200
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]
Cl3ErH12O18
MFCD00150232
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
erbium(3+);triperchlorate;hexahydrate

Specifications

14692-15-0
Liquid
MFCD00150232
Micible with water.
O.O.O.O.O.O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Er+3]
573.687
573.70 (465.61 Anhydrous)
Odorless
Erbium(III) perchlorate hexahydrate
Pink
Cl3ErH12O18
erbium iii perchlorate hexahydrate, erbium perchlorate-water 1/3/6, erbium iii perchlorate hexahydrate w/w in aqueous solution, reagent grade
JZRNHNTYRRYOCR-UHFFFAOYSA-K
erbium(3+);triperchlorate;hexahydrate
20038200
Reagent
Pink liquid
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H271-H315-H319-H335

missing translation for 'dotInformation' : DOT Class: 5.1, Packing Group: II

missing translation for 'einecsNumber' : 237-840-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only