missing translation for 'onlineSavingsMsg'
Learn More

Erbium pieces, distilled dendritic, 99.9% (REO), Thermo Scientific Chemicals

Product Code. 11381599
Click to view available options
Quantity:
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-52-0
Molecular Formula Er
Molecular Weight (g/mol) 167.26
MDL Number MFCD00010987
InChI Key UYAHIZSMUZPPFV-UHFFFAOYSA-N
Synonym unii-77b218d3ye, foil, iii ion, erbio, pieces, powder, chips, ingot, er3, pieces, distilled dendritic
PubChem CID 23980
ChEBI CHEBI:33379
IUPAC Name erbium
SMILES [Er]

Specifications

Color Silver
UN Number UN3178
Quantity 25 g
Percent Purity 99.9%
Sensitivity Moisture sensitive
Form Pieces
Assay (REO)
Chemical Name or Material Erbium pieces, Distilled dendritic

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.