missing translation for 'onlineSavingsMsg'
Learn More

Eriochrome Blue-Black R, Pure, C.I.15705, Metal Indicator, Fisher Chemical

Product Code. 10356420
Click to view available options
Quantity:
25 g
Packaging:
Glass Jar
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

MDL Number 4069
Formula Weight 416.39g/mol
Packaging Glass Jar
Color Black
pH 9.4
Quantity 25 g
Physical Form Powder
compliance-icons
Product Identifier
  • Calcon]TS
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.