Learn More
Thermo Scientific Chemicals Erioglaucine disodium salt, pure
35.18 € - 109.55 €
Chemical Identifiers
CAS | 3844-45-9 |
---|---|
Molecular Formula | C37H34N2Na2O9S3 |
Molecular Weight (g/mol) | 792.84 |
MDL Number | MFCD00012141 |
InChI Key | SGHZXLIDFTYFHQ-UHFFFAOYSA-L |
Synonym | brilliant blue fcf, acid blue 9, erioglaucine disodium salt, brilliant blue, FD&C blue no. 1, erioglaucine, alphazurine fg, FD&C blue no.1, fenazo blue xi, japan blue 1 |
PubChem CID | 19700 |
ChEBI | CHEBI:82411 |
IUPAC Name | disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate |
SMILES | CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10023110
|
Thermo Scientific Chemicals
229730050 |
5 g | Glass bottle |
35.18 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10562821
|
Thermo Scientific Chemicals
229730250 |
25 g | Glass bottle |
109.55 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3844-45-9 | |
792.84 | |
SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
19700 | |
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate |
C37H34N2Na2O9S3 | |
MFCD00012141 | |
brilliant blue fcf, acid blue 9, erioglaucine disodium salt, brilliant blue, FD&C blue no. 1, erioglaucine, alphazurine fg, FD&C blue no.1, fenazo blue xi, japan blue 1 | |
CHEBI:82411 | |
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] |
Specifications
3844-45-9 | |
283.0°C | |
10% max. (105°C) | |
C37H34N2Na2O9S3 | |
14, 855 | |
brilliant blue fcf, acid blue 9, erioglaucine disodium salt, brilliant blue, FD&C blue no. 1, erioglaucine, alphazurine fg, FD&C blue no.1, fenazo blue xi, japan blue 1 | |
SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulfonate | |
19700 | |
792.84 | |
Fine Crystalline Powder |
1000 (at lambda min.) | |
Purple | |
Authentic | |
MFCD00012141 | |
15, 1379 | |
Solubility in water: soluble. Other solubilities: soluble in ethanol, insoluble in vegetable oils | |
CCN(CC1=CC(=CC=C1)S(=O)(=O)[O-])C2=CC=C(C=C2)C(=C3C=CC(=[N+](CC)CC4=CC(=CC=C4)S(=O)(=O)[O-])C=C3)C5=CC=CC=C5S(=O)(=O)[O-].[Na+].[Na+] | |
792.84 | |
CHEBI:82411 | |
Pure | |
Erioglaucine disodium salt |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 223-339-8
missing translation for 'rtecsNumber' : BQ4725000
missing translation for 'tsca' : TSCA
RUO – Research Use Only