Learn More
Novus Biologicals™ ERK2 Knockout A549 Cell Lysate
Cell Lysate
Brand:  Novus Biologicals™ NBP3-18632
Product Code. 18602865
  • 637.00 € / 100µg
Explore more special offers
Specifications
P28482, P28482, P28482, P28482
100 μg
Human
Angiogenesis, Apoptosis, Breast Cancer, Cancer, Diabetes Research, Hypoxia, Phospho Specific, Protein Kinase, Signal Transduction, Tumor Suppressors
EC 2.7.11, EC 2.7.11.24, ERK, ERK-2, ERK2MAP kinase isoform p42, ERT1, Extracellular signal-regulated kinase 2, MAP kinase 1, MAP kinase 2, MAPK 1, MAPK2, mitogen-activated protein kinase 1, Mitogen-activated protein kinase 2, p38, p40, p41, p41mapk, p42-MAPK, P42MAPK, PRKM1MAPK 2, PRKM2, protein tyrosine kinase ERK2
This product has been tested by SDS-PAGE and western blot and is suitable for use in Western blot, ELISA, Immunoprecipitation and ChIP. No detection of expected band at ~44 kDa is observed in MAPK1 (ERK2) knockout A549 when compared with unmodified A549 cell lysates by Western blot.
Multi-step
Store at -70°C. Avoid freeze/thaw cycles.
ERK2
MAPK1