Learn More
Heidolph™ Erlenmeyer Attachment
For use with Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
Brand:  Heidolph™ 549-75000-00
 View more versions of this product
Product Code. 10408922
  • 205.00 € / Each
Estimated Shipment: 11-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Attachments for up to 22 Erlenmeyer flasks (25, 50, 100, 250, 500 or 1000 mL)
Specifications
Specifications
250 mL Erlenmeyer flasks
Erlenmeyer Attachment
Unimax 1010, Duomax 1030, Polymax 1040 and Promax 1020
5