Learn More
Erlenmeyer Flask for ML1 Microscale kit
Designed for organic chemistry experiments.
Brand:  Quickfit ML/FE25
 View more versions of this product
Product Code. 15778027
  • 399.00 € / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Designed for organic chemistry experiments using milligram quantities of reagents/li>
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 mL
Erlenmeyer flask