missing translation for 'onlineSavingsMsg'
Learn More

Lenz Laborglasintrumente Erlenmeyer Flasks, Narrow Neck

Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.

Brand:  Lenz Laborglasintrumente 07020052

 View more versions of this product

Product Code. 10000421

  • 130.38 € / Pack of 10
Estimated Shipment: 22-10-2024
to see stock.
Add to basket
This item is not returnable. View return policy

Description

Description

  • Acc. to DIN ISO 1773
  • With beaded rim
  • With graduation
  • Material: DURAN™
Specifications

Specifications

34 mm
Duran Glass
10/Pk.
300 mL
156 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers