Learn More
Ertugliflozin, MedChemExpress
MedChemExpress Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Brand:  Medchem Express HY-15461
Encompass_Preferred
 View more versions of this product
Product Code. 16422637
  • 167.40 € / 50mg
Explore more special offers
Description
Description
Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus.
Specifications
Specifications
Ertugliflozin
White
C22H25ClO7
PF-04971729
ClC1=CC=C([C@]23O[C@@](CO)(CO3)[C@@H](O)[C@H](O)[C@H]2O)C=C1CC4=CC=C(OCC)C=C4
436.88
Research
Metabolism-sugar/lipid metabolism
1210344-57-2
Research
50 mg
DMSO : 250 mg/mL (572.24 mM; Need ultrasonic)
436.88
98.82%
Solid