missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ ES 7X™-PF Cleaning Solution
ES 7X™-PF Cleaning Solution is a Phosphate Free detergent, specifically developed for safe and effective cleaning of laboratory instruments and glassware.
73.80 € - 2140.00 €
Product Code | Brand | Volume (English) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (English) | Price | Quantity & Availability | |||||
10019520
|
MP Biomedicals™
097667121 |
1.32 |
246.00 €
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409585
|
MP Biomedicals™
097667194 |
1 gal. |
239.00 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11419585
|
MP Biomedicals™
097667195 |
5 gal. | N/A | ||||||
17104185
|
MP Biomedicals™
097667195 |
5 gal. |
225.00 €
18.92 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11499575
|
MP Biomedicals™
097667193 |
1 gal. |
73.80 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429585
|
MP Biomedicals™
097667198 |
55 gal. | N/A | ||||||
17114185
|
MP Biomedicals™
097667198 |
55 gal. |
2140.00 €
208.19 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Environmentally safe and phosphate free.
- Developed to provide safe and effective cleaning of laboratory instruments and glassware including test tubes, petri-dishes, flasks, microscopy slides, pathology equipment and surgical instruments
- Several convenient package sizes are available
- Sold in concentrated form so the customer can prepare the appropriate dilution as needed
- Completely drains from glassware in seconds
- Eliminates residue leaving no interfering fluorescence
- pH neutral, and no need for pH adjustment at any concentration
- No etch to glass or plastic labware at any concentration
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ES-7X Cleaning Solution
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Dioctyl Sulfosuccinate Sodium Salt, 2-(2-butoxyethoxy)ethanol
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title