missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ES Series Combination Lab Refrigerator/Freezer, 230V

Use space in small work areas with the Thermo Scientific™ ES Series Combination Lab Refrigerator/Freezer, designed for routine sample protection.

Brand:  Thermo Scientific™ 263C-AEV-TS

1809.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 12673176

  • 2120.55 € / Each
Estimated Shipment: 03-02-2025
to see stock.
Explore more special offers
Add to basket

You may only order up to the available stock. Where an alternative item has been identified it will be shown below
This item is not returnable. View return policy

Description

Description

Effectively utilize space in small work areas with the Thermo Scientific™ ES Series Combination Lab Refrigerator/Freezer, designed for routine sample protection.

Features:

  • Integrated controller
  • Digital display
  • High and low temperature alarms
  • Standard door locks
  • Low energy consumption
  • Access ports
  • CE marked

Recommended for:

General-purpose lab storage

Note:

Available in 50Hz regions only; not available in North America or other 60Hz regions.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Combination General Purpose Refrigerator/Freezer
5.615 cu. ft.
159 L
1°C to 11°C
3 (Refrigerator), 1 (Freezer)
Alarm
13
Warranty varies by country, please contact us for details
16.3 in.
41.5 cm
66 in.
28.5 in.
67 cm
24 in.
21.4 in.
18 in.
40.8 cm
230 V
50 Hz
95 kg
3.849 cu. ft.
109 L
-12°C to -20°C
2
Manual
R600A
CE
24 x 21.4 x 66 in. (60 x 54.5 x 168 cm)
17.7 in.
45 cm
26.3 in.
168 cm
72.5 cm
60 cm
16 in.
54.5 cm
45.7 cm
230 V 50 Hz
209.5 lb.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.