Learn More
Notrax ESD Earth Bonding Plug
Product Code. 17741918
Click to view available options
:
Earth Bonding Plug; Color: Yellow
Product Code. 17741918
Brand: Notrax 054SEB0100
Only null left
Add to basket
Add to basket
Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment.
  • Specialized line of ESD accessories to ensure a static free workplace and protect employees and sensitive electrical equipment
  • Use these accessories to connect table work surface, employees and conductive material to a common grounding point
  • This creates a balance between all items and personnel to ensure static electricity will flow to ground in a controlled manner
  • For use in ESD working zones to protect electrostatic sensitive components
  • Connects grounding cords to ground
  • 1 Meg Resistor, 3 x 10 mm male studs, allowing multiple cord connections per plug
  • Available in Continental or UK version
Specifications
Color Yellow
Product Type Bounding Plug
Material Wire
Connector Types EU Plug Connection