Learn More
Cole-Parmer™ ESD Safe Plastic Tweezer with Flat, Duckbill Tips
ESD-safe tweezers, reinforced with carbon fiber, offer excellent wear and abrasion resistance. These tweezers are ideal for handling sensitive materials such as electronic components, micro-mechanical parts, glass, and ceramic substrates.
Brand:  Cole-Parmer™ 07387-17
Product Code. 15657418
  • 9.75 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • ESD-safe
  • Reinforced with 30% carbon fiber
  • Wear and abrasion resistant
  • Rigind, high tensile and flexural strength
  • Flat, duckbill tips
  • Handle sensitive electronic components safely
Specifications
Specifications
Flat, Duckbill Tips
ESD-safe plastic and carbon fiber
Sensitive materials such as electronic components, micro-mechanical parts, glass, and ceramic substrates
Non-sterile
Flat, Duckbill Tips
11.5 cm
ESD-safe plastic tweezers