missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Essential 8™ Medium

Essential 8™ Medium

Brand:  Gibco™ A1517001

294.21 EUR valid until 2025-12-31
Use promo code "24342" to get your promotional price.



Product Code. 15190617

  • 348.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Essential 8™ Medium is a xeno-free and feeder-free medium specially formulated for the growth and expansion of human pluripotent stem cells (PSCs). Originally developed by Guokai Chen et al. in the laboratory of James Thomson (published as 'E8') and validated by Cellular Dynamics International, Essential 8™ Medium has been extensively tested and proved to maintain pluripotency in multiple iPSC lines. In addition, Essential 8™ Medium has been used to scale up production of iPSCs and shown to support iPSC growth for >50 passages without any signs of karyotypic abnormalities, along with maintaining the ability of iPSCs to differentiate into all three germ line lineages.

Features of Essential 8™ Medium include:

  • Consistent—reduced variability compared to existing feeder-free culture media
  • Robust—reliable and robust cultures with a xeno-free, cGMP, 8-component medium
  • Cost effective—economical and scalable PSC culture compared to other feeder-free media

    Reduced Variability
    Essential 8™ Medium is xeno-free and contains only the 8 essential components needed for stem cell culture. Unlike other media that contain over 20 highly variable ingredients, Essential 8™ Medium is produced under cGMP and has an optimized formulation and growth factor levels to help ensure maximum cell health, pluripotency, and growth, with minimal variability.

    Reliable and Robust Cultures
    Essential 8™ Medium has been show to support pluripotent stem cell growth and provide cultures with superior morphology and growth kinetics compared to other feeder systems.

    Cost Effective
    Essential 8™ Medium is provided in a convenient two-component kit (500 ml basal medium & 10 ml supplement), and when used with vitronectin (VTN-N), provides a cost effective, defined system for feeder-free culture of human pluripotent stem cells (PSCs).

    Essential 8™ Medium is commercialized in partnership with Cellular Dynamics International.
    TRUSTED_SUSTAINABILITY
  • Specifications

    Specifications

    Embryonic Stem Cells (ESCs), Induced Pluripotent Stem Cells (iPSCs) 
    Liquid
    Serum-free
    cGMP for medical devices, 21 CFR Part 820 and ISO 13485
    500 mL
    Human
    Feeder-free Stem Cell Culture (Human, iPS - Induced Pluripotent Stem, Embryonic)
    Xeno-free
    Stem Cell Media
    Phenol Red
    Essential 8™
    Supplement - dry ice, Basal - ambient
    • 500 mL basal medium (store at 2—8°C and protect from light)
    • 10 mL supplement (store at -5 to -20°C and protect from light)
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling
    Show More
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    Product Content Correction

    Your input is important to us. Please complete this form to provide feedback related to the content on this product.

    Product Title
    Gibco™ Essential 8™ Medium > 500mL

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

    For Research Use Only. Not for use in diagnostic procedures.