Learn More
Estradiol 3-sulfamate, MedChemExpress
MedChemExpress Estradiol 3-sulfamate (BLE 00084; E2MATE; ES-J 995) is a potent, long-acting, and orally active steroid sulfatase inhibitor; inhibits estrone sulfatase with an IC50 of 251 nM and a Ki of 133 nM.
Brand:  Medchem Express HY-U00112
Encompass_Preferred
 View more versions of this product
Product Code. 16480169
  • 803.25 € / 10mg
Explore more special offers
Description
Description
Estradiol 3-sulfamate (BLE 00084
Specifications
Specifications
Estradiol 3-sulfamate
White
C18H25NO4S
BLE 00084 E2MATE ES-J 995
C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(OS(N)(=O)=O)=C3)([H])[C@]4([H])CC1
351.46
Research
Cancer-programmed cell death
172377-52-5
Research
10 mg
DMSO : 62.5 mg/mL (177.83 mM; Need ultrasonic)
351.46
99.5%
Solid