Learn More
Estradiol benzoate, MedChemExpress
MedChemExpress Estradiol Benzoate (β-Estradiol 3-benzoate), a prodrug of estradiol, acts as a steroid sex hormone. It exhibits mild anabolic and metabolic properties, and increases blood coagulability.
Brand:  Medchem Express HY-B1192
Encompass_Preferred
Product Code. 16442319
  • 91.80 € / 1g
Explore more special offers
Description
Description
Estradiol Benzoate (β-Estradiol 3-benzoate), a prodrug of estradiol, acts as a steroid sex hormone. It exhibits mild anabolic and metabolic properties, and increases blood coagulability.
Specifications
Specifications
Estradiol benzoate
White
C25H28O3
β-Estradiol 3-benzoate 17β-Estradiol 3-benzoate
C[C@@]12[C@@H](O)CC[C@@]1([H])[C@]3([H])CCC4=C(C=CC(OC(C5=CC=CC=C5)=O)=C4)[C@@]3([H])CC2
376.49
Research
Cancer-programmed cell death
50-50-0
Research
1 g
DMSO : 100 mg/mL (265.61 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
376.49
98.04%
Solid