Learn More
Estradiol, MedChemExpress
MedChemExpress Estradiol (β-Estradiol) is a steroid hormone and the major female sex hormone. Estradiol can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. Estradiol can be used for the research of cancers, neurodegenerative diseases and neural tissue engineering.
Brand:  Medchem Express HY-B0141
Encompass_Preferred
Product Code. 16462059
  • 91.80 € / 1g
Explore more special offers
Description
Description
Estradiol (β-Estradiol) is a steroid hormone and the major female sex hormone. Estradiol can up-regulate the expression of neural markers of human endometrial stem cells (hEnSCs) and promote their neural differentiation. Estradiol can be used for the research of cancers, neurodegenerative diseases and neural tissue engineering.
Specifications
Specifications
Estradiol
White
C18H24O2
β-Estradiol E2 17β-Estradiol 17β-Oestradiol
C[C@@]1([C@H]2O)[C@](CC2)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1
272.38
Research
Cancer-programmed cell death
50-28-2
Research
1 g
DMSO : ≥ 50 mg/mL (183.57 mM) ∣Ethanol : 20 mg/mL (73.43 mM; Need ultrasonic)
272.38
99.09%
Solid