missing translation for 'onlineSavingsMsg'
Learn More

β-Estradiol, Tocris Bioscience™

Product Code. 18712704
Click to view available options
Quantity:
100 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Endogenous ER agonist

β-Estradiol is an endogenous estrogen receptor (ER) agonist (Ki values are 0.12 and 0.13 nM for ERα and ERβrespectively). Also high affinity ligand at membrane estrogen GPR30 receptors. α-Estradiol also available.

Specifications

CAS 50-28-2
Molecular Formula C18H24O2
Quantity 100 mg
Synonym estradiol, beta-estradiol, 17beta-estradiol, oestradiol, dihydrofolliculin, estrace, ovocyclin, progynon, dihydrotheelin, dihydroxyestrin
InChI Key VOXZDWNPVJITMN-ZBRFXRBCSA-N
SMILES CC12CCC3C(C1CCC2O)CCC4=C3C=CC(=C4)O
IUPAC Name (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
Molecular Weight (g/mol) 272.388
PubChem CID 5757
ChEBI CHEBI:16469
Formula Weight 272.38
Percent Purity >99%
Chemical Name or Material β-Estradiol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'