Learn More
Estramustine phosphate sodium, MedChemExpress
MedChemExpress Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.
Brand: Medchem Express HY-13627
This item is not returnable.
View return policy
Description
Estramustine phosphate sodium, an estradiol analog, is an orally active antimicrotubule chemotherapy agent. Estramustine phosphate sodium depolymerises microtubules by binding to microtubule associated proteins (MAPs) and/or to tubulin. Estramustine phosphate sodium can interfere mitosis, trigger cell death and induce apoptosis, which can be used for the research of cancer like prostate cancer.Specifications
Estramustine phosphate sodium | |
White | |
C23H30Cl2NNa2O6P | |
H2O : 62.5 mg/mL (110.75 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (8.86 mM; Need ultrasonic) | |
564.35 | |
96.79% | |
Solid |
52205-73-9 | |
Research | |
100 mg | |
O=C(N(CCCl)CCCl)OC1=CC2=C(C=C1)[C@@]3([H])CC[C@@]4(C)[C@](CC[C@@H]4OP(O[Na])(O[Na])=O)([H])[C@]3([H])CC2 | |
564.35 | |
Research | |
Cancer-programmed cell death |
Research purposes only