missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estriol, MedChemExpress
MedChemExpress Estriol is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells.
Brand: Medchem Express HY-B0412
This item is not returnable.
View return policy
Description
Estriol is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells.Specifications
Estriol | |
White | |
C18H24O3 | |
Oestriol | |
C[C@@]1([C@H]2O)[C@](C[C@H]2O)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1 | |
288.38 | |
Research | |
Cancer-programmed cell death |
50-27-1 | |
Research | |
500 mg | |
DMSO : 250 mg/mL (866.91 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.47 mM; ultrasonic and warming and heat to 80°C) | |
288.38 | |
98.0% | |
Solid |
Research purposes only