missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Estriol, MedChemExpress
MedChemExpress Estriol is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells.
Brand: Medchem Express HY-B0412
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
Estriol is an antagonist of the G-protein coupled estrogen receptor in estrogen receptor-negative breast cancer cells.Specifications
Estriol | |
White | |
C18H24O3 | |
Oestriol | |
C[C@@]1([C@H]2O)[C@](C[C@H]2O)([H])[C@@](CCC3=C4C=CC(O)=C3)([H])[C@]4([H])CC1 | |
288.38 | |
Research | |
Cancer-programmed cell death |
50-27-1 | |
Research | |
100 mg | |
DMSO : 250 mg/mL (866.91 mM; Need ultrasonic) ∣H2O : 1 mg/mL (3.47 mM; ultrasonic and warming and heat to 80°C) | |
288.38 | |
98.0% | |
Solid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Research purposes only
Spot an opportunity for improvement?Share a Content Correction