missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva ETFE Ferrule
Ferrules used for chromatography tubing. Cytiva Lifescience™ ETFE Ferrule is made from ethylenetetra fluoroethylene and comes with different outside diameters.
108.58 € - 121.00 €
Specifications
For Use With (Equipment) | Tubing |
---|---|
Material | ETFE |
Type | Ferrule |
Quantity | 10 Pack |
Product Code | Brand | Diameter (English) Outside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (English) Outside | Price | Quantity & Availability | |||||
11360252
|
Cytiva
18-1121-18 |
1/8 in. |
113.00 €
Each |
In Stock
Log in to see stock. |
|||||
11300262
|
Cytiva
18-1127-06 |
1/16 in. |
121.00 €
Each |
In Stock
Log in to see stock. |
|||||
11360052
|
Cytiva
18-1112-48 |
3/16 in. |
108.58 €
Each |
In Stock
Log in to see stock. |
|||||
Description
- Ferrules
- Chromatography accessory
- Used for: protecting the end of tubing
- Packsize: 10 Connections
- Ethylenetetra fluoroethylene (ETFE)
Specifications
Tubing | |
ETFE | |
Ferrule | |
10 Pack |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title