missing translation for 'onlineSavingsMsg'
Learn More

Cole-Parmer™ Square Tipped Autoclavable Tezfel (ETFE) Tongs

Get a grip on your samples with these Square Tipped Autoclavable Tezfel (ETFE) Tongs.

Brand:  Cole-Parmer™ 06442-00

Product Code. 11755279

  • 55.45 € / Each
Estimated Shipment: 27-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Autoclavable.
  • Tefzel ETFE tongs measures 10 in. long and has 3/4 in. wide squared tips.
  • Use to reach into deep tanks.
Specifications

Specifications

Use to reach into deep tanks
1 Ea.
254 mm
Tongs
10 in.
Square
ETFE
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cole-Parmer™ Square Tipped Autoclavable Tezfel (ETFE) Tongs > Square Tipped Autoclavable Tongs; Material: ETFE

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.