Learn More
IKA™ ETH SER 4 Ethernet-Serial Converter
With the ETH SER 4 Ethernet-Serial Converter, up to four laboratory devices can be controlled or read out via ethernet.
Brand:  IKA™ 0020024285
Product Code. 17287499
  • 2245.00 € / Each
Estimated Shipment: 05-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Up to 4 lab units can be controlled through the ethernet with the PC 4.1 RS-232 server
  • Server supports 4 RS-232 ports with a 10/100 mbps ethernet interface by TCP/IP
  • Server can be set-up through the ethernet and works as a transparent serial COM-Port without restrictions of platform and distance
Specifications
Specifications
Ethernet-serial Converter
RS-232 (4 Ports)