missing translation for 'onlineSavingsMsg'
Learn More

Ethane, SPEX CertiPrep™

Product Code. 12919234
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Ethane74-84-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 74-84-0, 67-56-1
Packaging 2mL amber prescored glass ampule
CAS Min % 0.1, 99.9
Quantity 1 mL
CAS Max % 0.1, 99.9
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Molecular Formula C2H6
MDL Number MFCD00009023
InChI Key OTMSDBZUPAUEDD-UHFFFAOYSA-N
SMILES CC
IUPAC Name ethane
Molecular Weight (g/mol) 30.07
PubChem CID 6324
ChEBI CHEBI:42266
Solvent or Matrix Methanol-P&T
Chemical Name or Material Ethane
Show More Show Less

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'