missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanediol, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Plastic container
Description
C2H6O2, CAS Number-107-21-1, ethylene alcohol, glycol alcohol, 2-hydroxyethanol, 1,2-ethanediol, glycol, fridex, monoethylene glycol, tescol, 1,2-dihydroxyethane, ethylene glycol, 2.5L, 196 deg.C, CHEBI:30742, Colorless, 62.06g/mol, LYCAIKOWRPUZTN-UHFFFAOYSA-N, ethane-1,2-diol, -13 deg.C, 2885
Specifications
Specifications
Melting Point | -13°C |
Color | Colorless |
pH | 5.5 to 7.5 |
Boiling Point | 196°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Vapor Density | 2.14 |
Formula Weight | 62.06g/mol |
Vapor Pressure | 0.12mmHg at 20°C |
Physical Form | Viscous Liquid |
Safety and Handling
Product Identifier
- Ethylene glycol
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity after repeated exposure Category 2
- H373-May cause damage to organs.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /A§
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'