missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 200 Proof (100%), USP/EP/ACS, Tax-Paid, Fisher Chemical™
A tax-paid 200 proof ethanol that meets USP, EP, and ACS testing specifications to encourage cross-regional collaboration and potentially reduce laboratory inventory for ethanol applications.
373.00 €
Chemical Identifiers
CAS | 64-17-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Description
- Meets USP, EP, and ACS testing specifications
- Consolidate your purchases into one product code and potentially reduce inventory
- Suitable for use as a cleaner, solvent, or reagent in various applications
- Packaged in laboratory-friendly sizes
- Pricing includes tax
- Combination of compendial test specifications enables use across regions
- Intended for research use only; not for human or animal diagnostic purposes
Chemical Identifiers
64-17-5 | |
46.069 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
C2H6O | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
Specifications
-114°C | |
64-17-5 | |
Liquid | |
C2H6O | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
ACS/EP/USP |
0.40 max. at 240nm (USP, EP), 0.30 max at 250-260nm (USP, EP), 0.10 max at 270-340nm (USP, EP) | |
78°C | |
0.001% max. (ACS) | |
0.2% max. (ACS) | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07 | |
Ethanol, 200 Proof |
Safety and Handling
Note: Products are tested to meet the analytical requirements of the noted grade (USP, Ph. EUR or otherwise). Product test results are available on the Certificate of Analysis or upon request for the specific product.
Products are for research use or further manufacturing. Products are not for direct administration to humans or animals. It is the responsibility of the final formulator and end-user to qualify, test and/or validate each product for its intended use as an ingredient in the end product and in accordance with applicable regulations (including without limitation WHO-recommended Hand Rub Formulations (Hand Sanitizers) and/or FDA Policy for Preparation of Certain Alcohol-based Hand Sanitizer Products, as updated). Thermo Fisher Scientific expressly disclaims all non-specified warranties, expressed or implied, including the implied warranties of merchantability and fitness for a particular use.
For Research Use Only