missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 70%, technical, denaturated with 5% methanol Product Code.: 13227993

Ethanol, 70%, technical, denaturated with 5% methanol

Product Code. 13227993
5 L, Plastic Drum
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 13227993

Brand: Thermo Scientific Acros 444200050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

CAS: 64-17-5 | C2H6O | 46.07 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ethanol, 70%, technical, denaturated 5L with 5% methanol Plastic Bottle Cas No. 64-17-5
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -114°C
CAS 64-17-5
Boiling Point 78°C
Quantity 5 L
Packaging Plastic Drum
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Percent Purity 70%
Chemical Name or Material Ethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466549 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number444200050
Lot NumberA0467519 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number444200050
Lot NumberA0459891 Certificate TypeCertificate of Analysis Date11/03/2024 Catalog Number444200050
Lot NumberA0454828 Certificate TypeCertificate of Analysis Date01/09/2023 Catalog Number444200010,444200050
Lot NumberA0451141 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number444200050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ethanol, 70% denaturated with 5% methanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H371-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P308-IF exposed or concerned:.

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 70%, technical, denaturated with 5% methanol >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.