missing translation for 'onlineSavingsMsg'
Learn More
Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical™ Product Code.: 10090100

Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical™

Product Code. 10090100
1 L, Amber glass bottle
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
Amber glass bottle
Metal can
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 10090100

Brand: Fisher Chemical E/0550DF/15

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
MDL Number 3568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236

Specifications

Packaging Amber glass bottle
Quantity 1 L
Molecular Formula C2H6O
Physical Form Liquid
Color Colorless
Melting Point -114°C
Boiling Point 78°C
Formula Weight 46.07g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2525705 Certificate TypeCertificate of Analysis Date05/02/2025 Catalog NumberE/0550DF/25,E/0550DF/15,E/0550DF/C17,E/0550DF/21,E/0550/08,E/0550DF/17,E/0550/17,E/0550/PC17
compliance-icons compliance-icons
Product Identifier
  • Ethanol (95% v/v)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol, 95% (v/v), Certified AR for Analysis, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.