missing translation for 'onlineSavingsMsg'
Learn More

Ethanol 95% (v/v), Extra Pure, SLR, Fisher Chemical™

50.95 € - 377.00 €

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
MDL Number 3568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 7
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10510153
View Documents
Fisher Chemical
E/0500DF/08
500 mL Amber Glass Bottle
53.50 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10681533
View Documents
Fisher Chemical
E/0500DF/15
1 L Amber Glass Bottle
50.95 €
1 L
Please to purchase this item. Need a web account? Register with us today!
10040602
View Documents
Fisher Chemical
E/0500DF/17
2.5 L Amber Glass Bottle
78.95 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10572143
View Documents
Fisher Chemical
E/0500DF/C17
2.5 L HDPE Bottle
215.00 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
12666697
View Documents
Fisher Chemical
E/0500DF/P17
2.5 L Plastic-coated Glass Bottle
73.55 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
10305752
View Documents
Fisher Chemical
E/0500DF/P21
5 L HDPE Bottle
220.00 €
5 L
Please to purchase this item. Need a web account? Register with us today!
10447492
View Documents
Fisher Chemical
E/0500DF/P24
10 L Plastic Container
377.00 €
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

64-17-5
3568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO
C2H6O
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol

Specifications

-114°C
Colorless
Liquid
3568
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.07g/mol
64-17-5
78°C
C2H6O
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CCO
46.069
CHEBI:16236
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Ethanol (95% v/v)

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.