missing translation for 'onlineSavingsMsg'
Learn More

Ethanol ≥ 96% (v/v), BAKER ANALYZED™ ACS, J.T. Baker™

Product Code. 10730322
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Exceeds ACS Specifications

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO

Specifications

Melting Point -114°C
Color Max. 10 (APHA)
Quantity 1 L
Assay Percent Range 95.1 to 96.9% (v/v)
Residue after Evaporation max. 0.001%
Titratable Acid max. 0.0005 meq/g
Titratable Base max. 0.0002 meq/g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.