missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 96% v/v, BAKER ANALYZED™, J.T. Baker™

Product Code. 15558454
Click to view available options
Quantity:
2.5 L
Packaging:
HDPE
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Exceeds ACS specifications to provide improved consistency and characterization of the reagent.

BAKER ANALYZED™ reagents are a grade of high quality chemicals for laboratory and specialized industrial use. Every BAKER ANALYZED™ Reagent label provides an Actual Lot Analysis as a guarantee of purity and consistency. Analytical methods used for these determinations are based on standard test procedures described in technical publications.

Specifications

Melting Point −114.5°C
CAS 64-17-5
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 78°C
Quantity 2.5 L
Explosion Limit Lower: 3.5 Vol %, Upper: 15 Vol %
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol
Packaging HDPE
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Molecular Formula C2H5OH
Titratable Acid Max. 0.0005 meq/g
Titratable Base Max. 0.0002 meq/g
Viscosity 1.2 mPas at 20°C
Molecular Weight (g/mol) 46.07
Percent Purity Min. 95.1 to 96.9% v/v
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons
Product Identifier
  • 2,5LT Ethanol, 96% (v/v) BAKER ANALYZED Reagent
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Supplemental information
  • MIXTURE LIST-Contains : ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.