missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, 99.5%, Extra Dry, absolute, AcroSeal™, Thermo Scientific Chemicals
Ethanol, >-99.5%, C2H6O, CAS Number-64-17-5, 7732-18-5, algrain, alcohol, ethyl hydroxide, ethyl hydrate, alcohol, ethyl alcohol, anhydrol, methylcarbinol, tecsol, grain alcohol, 100mL, CHEBI:16236, Colorless, 0.7900g/mL, 46.07, Extra Dry, Danger, GHS P Statement Highly flammable liquid and vapor.
56.60 € - 179.00 €
Chemical Identifiers
CAS | 64-17-5, 7732-18-5 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10617864
|
Thermo Scientific Acros
397691000 |
100 mL | AcroSeal™ Glass Bottle |
56.60 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10483195
|
Thermo Scientific Acros
397690010 |
1 L | AcroSeal™ Glass Bottle |
96.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10646134
|
Thermo Scientific Acros
397690025 |
2.5 L | AcroSeal™ Glass Bottle |
179.00 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-17-5, 7732-18-5 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
64-17-5, 7732-18-5 | |
0.7900g/mL | |
Liquid | |
0.001% max. | |
CH3CH2OH | |
1170 | |
15, 3814 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.07 | |
Extra Dry |
Colorless | |
Authentic | |
99.5% | |
C2H6O | |
MFCD00003568 | |
0.005% max. | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CCO | |
46.069 | |
CHEBI:16236 | |
≥99.5% | |
Ethanol, Extra Dry |
Safety and Handling
GHS P Statement Highly flammable liquid and vapor.
GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Danger
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Ethanol
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P337+P313-If eye irritation persists: Get medical advice/attention.
For Manufacturing and Laboratory Use. Not for beverage use