missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, 99.8%, technical, not denaturated, Thermo Scientific Chemicals

Product Code. 13237993
Click to view available options
Quantity:
5 L
Packaging:
Plastic Drum
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Melting Point -114.0°C
CAS 64-17-5
Density 0.7900 g/mL
Boiling Point 78.0°C
Quantity 5 L
Flash Point 12°C
Packaging Plastic Drum
Assay Percent Range 99.8%
Specific Gravity 0.79
Molecular Formula C2H6O
Linear Formula CH3CH2OH
Viscosity 1.2 mPa.s (20°C)
Merck Index 15, 3814
Solubility Information Solubility aq. soln.: completely soluble.
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Formula Weight 46.07
Grade Technical
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

For Manufacturing and Laboratory Use. Not for beverage use

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'