missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute (200 Proof), Molecular Biology Grade, Fisher BioReagents™

Product Code. 16688242
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
4 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Meets ACS Specifications

  • Meets ACS Specifications
  • For nucleic acid purification and precipitation
  • Ultrapure molecular biology grade ethanol is used for the purification and precipitation of biomolecules such as nucleic acids and proteins
  • It is also used in histology to prepare staining and destaining reagents and for dehydrating tissues prior to embedding
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 64-17-5
Molecular Formula C2H6O
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Spezifikation

Melting Point -114°C
Color Colorless
Boiling Point 78°C
Quantity 2.5 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Infrared Spectrum Conforms to reference
Physical Form Liquid
Assay Percent Range Assay: ≥99.5% (v/v)
Residue after Evaporation 0.001% max.
Titratable Acid 0.0005 mEq /g max.
Linear Formula C2H5OH
Titratable Base 0.0002 mEq /g max.
Water 0.2% max.
Merck Index 15, 3814
Molecular Weight (g/mol) 46.07
Formula Weight 46.07
Percent Purity ≥99.5%
Grade DNase-, RNase- and Protease-Free
Chemical Name or Material Ethanol, Absolute (200 Proof)
Mehr anzeigen Weniger anzeigen
compliance-icons compliance-icons
Produktidentifikator
  • Ethanol, for molecular biology
Signalwort
  • Danger
Gefahrenkategorie
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Gefahrenhinweis
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Sicherheitshinweis
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P370+P378-In case of fire: Use to extinguish.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Ergänzende Informationen
  • MIXTURE LIST-Contains : ethanol
Berichtigung von Produktinhalten

Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.

Name des Produkts

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt