missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Metal can
Plastic Coated Glass bottle
Plastic container
Description
C2H6O, CAS Number-64-17-5, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702, CCO

Specifications
Specifications
Packaging | Amber glass bottle |
Quantity | 1 L |
Physical Form | Liquid |
Color | Colorless |
pH | 7 |
Melting Point | -114°C |
Boiling Point | 78°C |
Formula Weight | 46.07g/mol |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
2526667 | Certificate of Analysis | 11/02/2025 | E/0650DF/25,E/0650DF/99X,E/0650DF/R17,E/0650DF/24,E/0650DF/15Z,E/0650DF/P17,E/0650DF/HD25,E/0650DF/15,E/0650DF/17Z,E/0650DF/21,E/0650DF/28,E/0650DF/17,E/0650DF/21Z,E/0650DF/08,E/0650/17,E/0650DF/C17,E/0650DF/C17Z,E/0650/15,E/0650/08,E/0650DF/25SS,E/0650/25,E/0650DF/PC15,E/0650/PC17 |
Safety and Handling


Product Identifier
- Ethanol 96%, technical, duty free
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H319-Causes serious eye irritation.
- H225-Highly flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- MIXTURE LIST-Contains : Ethanol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ethanol Absolute 99.8+%, Certified AR for Analysis, meets analytical specification of Ph.Eur., BP, Fisher Chemical™ >
Spot an opportunity for improvement?Share a Content Correction