missing translation for 'onlineSavingsMsg'
Learn More

Ethanol Absolute, 99.9%, BAKER ANALYZED™, J.T. Baker™

Product Code. p-7145019 Shop All J.T.Baker Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
HDPE Bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 15578454

Brand: JT Baker 8025.1000PE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Exceeds ACS specifications to provide improved consistency and characterization of the reagent. J.T. Baker™ Ethanol Absolute, 99.9%, BAKER ANALYZED™ meets EPA method 130.2 for testing of water hardness and meets US FDA requirements for use in the pharmaceutical industry.

BAKER ANALYZED™ reagents are a grade of high quality chemicals for laboratory and specialized industrial use. Every BAKER ANALYZED™ Reagent label provides an Actual Lot Analysis as a guarantee of purity and consistency. Analytical methods used for these determinations are based on standard test procedures described in technical publications.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07

Specifications

Melting Point −114.5°C
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 78°C
Quantity 1 L
Explosion Limit Lower: 3.5 Vol %, Upper: 15 Vol %
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol
Packaging HDPE Bottle
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Titratable Acid Max. 0.0005 meq/g
Titratable Base Max. 0.0002 meq/g
Viscosity 1.2 mPas at 20°C
Water Max. 0.2%
Solubility Information 1g/L in water at 20°C
Percent Purity Min. 99.9% v/v
Chemical Name or Material Ethanol, Absolute
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol Absolute
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.