missing translation for 'onlineSavingsMsg'
Learn More

Ethanol absolute, BAKER ANALYZED™, HPLC, J.T. Baker™

Product Code. 10017680
1 L
Click to view available options
1 L
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

For HPLC, analytical reagent

  • Ultraviolet Absorbance (1.00-cm path vs water):
    • at 210 nm max. 0.7
    • at 240 nm max. 0.1
    • at 260 nm max. 0.01

Specifications

Melting Point -117°C
CAS 64-17-5
Density 0.7895 g/cm³ (20°C)
Boiling Point 78.3°C (1013 hPa)
Quantity 1 L
Flash Point 12°C
Packaging Glass Bottle
Assay Percent Range min. 99.5%
Residue after Evaporation max. 0.001%
Molecular Formula C2H6O
MDL Number MFCD00003568
Water max. 0.2%
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.07
Show More Show Less
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol absolute, BAKER ANALYZED™, HPLC, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.