missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, Absolute, BAKER ANALYZED, J.T. Baker™

Product Code. 15598454
Click to view available options
Quantity:
5 L
Packaging:
HDPE
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

High quality grade chemical that exceeds ACS specifications for laboratory and specialized industrial use.

  • Exceeds the requirements set forth by the American Chemical Society (ACS)
  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Specifications

Melting Point -114.5°C
CAS 64-17-5
Color Colorless
Density 0.79g/cm3 at 20°C
Boiling Point 78°C
Quantity 5 L
Explosion Limit 3.5 - 15 Vol %
Flash Point 12°C
Ignition Point 425°C
Odor Alcohol-like
Packaging HDPE
Physical Form Liquid
Residue after Evaporation Max. 0.001%
Molecular Formula C2H6O
Titratable Acid Max. 0.0005meq/g
Linear Formula C2H5OH
Titratable Base Max. 0.0002meq/g
Viscosity 1.2 mPas at 20°C
Water Max. 0.2%
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
SMILES CCO
IUPAC Name ethanol
Molecular Weight (g/mol) 46.069
PubChem CID 702
ChEBI CHEBI:16236
Formula Weight 46.07g/mol
Percent Purity Min. 99.9%
Grade Exceeds ACS specifications
Chemical Name or Material Ethanol, Absolute
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol Absolute
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P235-Store in a well-ventilated place. Keep cool.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'