missing translation for 'onlineSavingsMsg'
Learn More
Ethanol Absolute, for HPLC, Fisher Chemical™ Product Code.: 12438760

Ethanol Absolute, for HPLC, Fisher Chemical™

Product Code. 12438760
2.5 L, Plastic Bottle
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Amber Glass Bottle
Plastic Bottle
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Product Code. 12438760

Brand: Fisher Chemical E/0665DF/P17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

C2H6O, CAS Number-64-17-5, alcohol, algrain, alcohol, anhydrol, ethyl alcohol, ethyl hydrate, ethyl hydroxide, grain alcohol, methylcarbinol, tecsol, 2.5L, 78 deg.C, CHEBI:16236, Colorless, 46.07g/mol, LFQSCWFLJHTTHZ-UHFFFAOYSA-N, ethanol, -114 deg.C, 3568, 46.069, Amber glass bottle, Liquid, 702

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
MDL Number 3568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

pH 7
Melting Point -114°C
Color Colorless
Boiling Point 78°C
Quantity 2.5 L
Packaging Plastic Bottle
Physical Form Liquid
Synonym ethyl alcohol
Molecular Weight (g/mol) 46.07
Formula Weight 46.07g/mol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528690 Certificate TypeCertificate of Analysis Date26/02/2025 Catalog NumberE/0665DF/17Z,E/0665DF/15X,E/0665DF/17,E/0665DF/27Z,E/0665DF/17X,E/0665DF/15,E/0665DF/17CN,E/0665/17,E/0665/15,E/0665DF/P17
Lot Number2526667 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberE/0665DF/17Z,E/0665DF/17,E/0665DF/15X,E/0665DF/27Z,E/0665DF/17X,E/0665DF/15,E/0665DF/17CN,E/0665/17,E/0665/15,E/0665DF/P17
compliance-icons compliance-icons
Product Identifier
  • Ethanol, absolute
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ethanol Absolute, for HPLC, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.