missing translation for 'onlineSavingsMsg'
Learn More

Ethanol (Untaxed), Puriss. p.a., absolute, ≥99.8% (GC), for license holders only, Honeywell Riedel-de Haën™

Product Code. 15603720
Click to view available options
Quantity:
2.5 L
5 L
10 L
30 kg
4 x 2.5 L
6 x 1 L
160 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly-coated Bottle
Stainless Steel Can
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO

Specifications

Density 0.789g/mL at 25°C
Quantity 10 L
Explosion Limit 19% (60°F)
Flash Point 14°C (57.2°F)
Ignition Point 683°F
Packaging Stainless Steel Can
Linear Formula CH3CH2OH
Vapor Pressure 44.6mmHg (20°C)
UN Number 1170
Beilstein 1718733
Formula Weight 46.07g/mol
Percent Purity ≥99.8% (GC)
Grade puriss. p.a.
Chemical Name or Material Ethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'