missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol, Anhydrous, Honeywell™ Riedel-de-Haën™
Click to view available options
Quantity:
1 L
5 L
Specifications
Specifications
Density | 0.789g/mL at 25°C |
Quantity | 1 L |
Explosion Limit | 19% (60°F) |
Flash Point | 14°C (57.2°F) |
Ignition Point | 683°F |
Linear Formula | CH3CH2OH |
Vapor Pressure | 44.6mmHg (20°C) |
UN Number | UN1170 |
Beilstein | 1718733 |
Formula Weight | 46.07g/mol |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction