missing translation for 'onlineSavingsMsg'
Learn More

Ethanol, CHROMASOLV™ Gradient Grade, Honeywell Riedel-de Haën™

for HPLC, gradient grade,≥99.8%, for license holders only

71.40 € - 160.69 €

Chemical Identifiers

CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.069
MDL Number MFCD00003568
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
Synonym ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
PubChem CID 702
ChEBI CHEBI:16236
IUPAC Name ethanol
SMILES CCO
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15610600
View Documents
Honeywell-Chromasolv
02854-1L
1 L
71.40 €
1 L
Please to purchase this item. Need a web account? Register with us today!
15620600
View Documents
Honeywell-Chromasolv
02854-2.5L
2.5 L
160.69 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
Specifications

Chemical Identifiers

64-17-5
46.069
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
702
ethanol
C2H6O
MFCD00003568
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
CHEBI:16236
CCO

Specifications

64-17-5
19% (60°F)
683°F
CH3CH2OH
44.6mmHg (20°C)
1718733
Completely souble in water
CCO
46.069
CHEBI:16236
≥99.8%
Ethanol
0.789g/mL at 25°C
14°C (57.2°F)
C2H6O
MFCD00003568
1170
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol
LFQSCWFLJHTTHZ-UHFFFAOYSA-N
ethanol
702
46.07g/mol
CHROMASOLV™ Gradient
Videos
Safety and Handling

Safety and Handling

P210-P305 + P351 + P338-P370 + P378-P403 + P235

H225-H319

missing translation for 'einecsNumber' : 200-578-6

missing translation for 'rtecsNumber' : KQ6300000

Product Identifier
  • Ethanol

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : ethanol

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.