missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Ethanol, CHROMASOLV™, Honeywell Riedel-de Haën™
Klik for at se tilgængelige muligheder
Quantity:
250 mL
1 L
2.5 L
4 x 2.5 L
Pakningsstørrelse:
1 L
2.5 L
250mL
Pack of 4
Tekniske data
Tekniske data
| Density | 0.789g/mL at 25°C |
| Quantity | 4 x 2.5 L |
| Explosion Limit | 19% (60°F) |
| Flash Point | 14°C (57.2°F) |
| Ignition Point | 683°F |
| Physical Form | Absolute Alcohol |
| Linear Formula | CH3CH2OH |
| Vapor Pressure | 44.6mmHg (20°C) |
| UN Number | 1170 |
| Beilstein | 1718733 |
| Vis mere |
Sikkerhed og håndtering
Produktidentifikator
- Ethanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion