Learn More
Ethanol, denatured, Spectrophotometric Grade, 90%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 64-17-5, 67-56-1, 67-63-0 |
---|---|
Molecular Formula | C2H6O |
Molecular Weight (g/mol) | 46.069 |
MDL Number | MFCD00003568 |
InChI Key | LFQSCWFLJHTTHZ-UHFFFAOYSA-N |
Synonym | ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol |
PubChem CID | 702 |
ChEBI | CHEBI:16236 |
IUPAC Name | ethanol |
SMILES | CCO |
Description
Ethanol, anhydrous, denatured is used in spectrophotometry and environmental testing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
64-17-5, 67-56-1, 67-63-0 | |
46.069 | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
702 | |
ethanol |
C2H6O | |
MFCD00003568 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
CHEBI:16236 | |
CCO |
Specifications
°C to 90°C | |
Colorless | |
0.789 | |
11°C (52°F) | |
Alcohol-like | |
1.361 | |
C2H6O | |
57 hPa (43mm Hg) at 20°C | |
1718733 | |
Miscible with water and most organic solvents. | |
CCO | |
46.069 | |
CHEBI:16236 | |
Spectrophotometric | |
Ethanol, denatured |
64-17-5, 67-56-1, 67-63-0 | |
Colorless liquid | |
78°C | |
455°C (851°F) | |
Liquid | |
90% | |
MFCD00003568 | |
1987 | |
ethyl alcohol, alcohol, methylcarbinol, grain alcohol, ethyl hydroxide, ethyl hydrate, algrain, alcohol, anhydrol, tecsol | |
LFQSCWFLJHTTHZ-UHFFFAOYSA-N | |
ethanol | |
702 | |
46.08 | |
Ethyl alcohol: 90.0%; Methanol: 5.0%; 2-Propanol: 5.0% |
Safety and Handling
Keep container tightly closed and in a well-ventilated place. Keep away from sources of ignition - No Smoking. Avoid contact with eyes. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P309+P311-P312-P330-P370+P378q-P501c
H225-H302+H332-H319-H336-H371
missing translation for 'dotInformation' : Hazard Class: 3, Packing Group: II
missing translation for 'einecsNumber' : 200-578-6
missing translation for 'rtecsNumber' : KQ6300000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
For Manufacturing and Laboratory Use. Not for beverage use
Your input is important to us. Please complete this form to provide feedback related to the content on this product.