Learn More
Gibco™ Ethanol (EtOH), 20%
Gibco 20% Ethanol (EtOH) is sterile-filtered, packaged in bioprocess containers, and stocked to be available when you need it. Sourcing 20% EtOH can streamline your workload for equipment sanitization and column storage, allowing you to focus on the key steps in your downstream processes.
614.00 € - 5670.00 €
Specifications
Chemical Name or Material | Ethyl Alcohol |
---|---|
Color | Clear |
Physical Form | Liquid |
Certifications/Compliance | GMP - 21 CFR 820, ISO 13485 |
Concentration | 20% |
Product Code | Brand | Packaging | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Packaging | Quantity | Price | Quantity & Availability | ||||
18034060
|
Gibco™
A5803201 |
Sample | 5 L |
614.00 €
5 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18034100
|
Gibco™
A5803202 |
Tote | 20 L |
1055.00 €
20 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
18004090
|
Gibco™
A5803203 |
Drum | 200 L |
5670.00 €
200 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Ethanol (EtOH) and bulk Water for Injection (WFI) are mixed to produce solutions that meet the most stringent quality control expectations in accordance with the pharmaceutical, biopharma, and diagnostic industries. Gibco 20% Ethanol is manufactured to cGMP standards at an ISO certified facility, promoting quality and convenience for every stage of a biologic’s pipeline.
Product features:
• Available in a range of pack sizes (5 L, 20 L, 200 L)
• Product specifications are well-defined, with the established solutions ready to order
• Manufactured to ISO and cGMP standards to meet the stringent expectations of highly regulated industries
• Readily accessible documentation for accelerated qualification
User benefits:
• Meets workflow needs for all stages of biologic development and manufacturing
• Streamlines in-house processes related to formulation, hydration, testing, and validation
• Mitigates space required for buffer preparation and storage
• Eases the time requirements dedicated for in-house buffer preparation
Specifications
Ethyl Alcohol | |
Liquid | |
20% | |
20% EtOH |
Clear | |
GMP - 21 CFR 820, ISO 13485 | |
0.2 μm Sterile-filtered |
Safety and Handling
missing translation for 'shelfLife' : 12 Months
missing translation for 'dotInformation' : DOT Transport Hazard Class 3, Packaging Group III
For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.