missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss., meets analytical spec. of BP, Ph. Eur., 96% (v/v), for license holders only, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
160 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly Bottle
Stainless Steel Can
Specifications
Specifications
Melting Point | -114°C (literature) |
CAS Max % | 100.0000% |
Density | 0.789 g/mL at 25 °C(lit.) |
Boiling Point | 78°C to 79°C |
Quantity | 1 L |
Flash Point | 12°C |
Packaging | Poly Bottle |
Linear Formula | NaHSO3 |
MDL Number | MFCD00003530 |
UN Number | UN1170 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'