missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), Puriss., meets analytical spec. of BP, Ph. Eur., 96% (v/v), for license holders only, Honeywell Riedel-de Haën™
![BIOPHARMA](/content/dam/fishersci/glyphs/sure_trace.png)
Click to view available options
Quantity:
1 L
2.5 L
5 L
30 kg
4 x 2.5 L
4 x 5 L
160 kg
Packaging:
Glass Bottle
Metal Drum
Plastic Bottle
Poly Bottle
Stainless Steel Can
Specifications
Specifications
Density | 0.789g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 19% (60°F) |
Flash Point | 14°C (57.2°F) |
Ignition Point | 683°F |
Packaging | Plastic Bottle |
Linear Formula | CH3CH2OH |
MDL Number | MFCD00003568 |
Vapor Pressure | 44.6mmHg (20°C) |
UN Number | UN1170 |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS02_Extremely_Flammable.png)
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Signal Word
- Danger
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction