missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Ethanol Safety Wash Bottle
Click to view available options
Capacity (Metric):
500 mL
Description
- Manufactured from yellow transparent PE-LD
- Contents label in German, English, French and Spanish
- Hazard symbols and hazard indications pursuant to the German Ordinance on Hazardous Substances
- 500 mL capacity
Specifications
Specifications
Capacity (Metric) | 500 mL |
Color | Translucent Yellow |
For Use With (Application) | Ethanol |
Includes | Screw cap, spray tube |
Material | LDPE |
Certifications/Compliance | German Ordinance on Hazardous Substances |
Product Type | Safety wash bottle |
Closure Type | Screw Cap |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction