missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethanol (Untaxed), CHROMASOLV™, absolute, for HPLC, sales not in Germany1, ≥99.8% (GC), for license holders only, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2.5 L-PC
4 x 2.5 L
6 x 1 L
Specifications
Specifications
Melting Point | -114°C |
Density | 0.789g/mL at 25°C |
Boiling Point | 78°C |
Quantity | 1 L |
Refractive Index | n20/D 1.3600 |
Linear Formula | CH3CH2OH |
Vapor Pressure | 44.6mmHg (20 °C) |
Beilstein | 1718733 |
Formula Weight | 46.07g/mol |
Percent Purity | ≥99.8% (GC) |
Show More |
Safety and Handling
Product Identifier
- Ethanol
- Danger
- Flammable liquid Category 2
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'